Would you like to log in again?
Self-Standing Centrifuge Tubes, Sterile
Supplier: United Scientific Supplies
P10424
470339-444EA 196.8 USD InStock
470339-444
Self-Standing Centrifuge Tubes, Sterile
Tubes Centrifuge Tubes
Sterilized self standing centrifuge tubes.
  • Pack of 500 self-standing polypropylene centrifuge tubes
  • Leak-proof, blue HDPE screw caps included
  • Silk-screen graduations and large white labeling area
  • 29.2O.D×117.3H mm
  • Maximum RCF: 9000
Polypropylene centrifuge tubes feature a flat bottom and 50 ml capacity. Leak-proof, blue HDPE screw caps included. Silk-screen graduations and large white marking area on each tube.
Autoclavable at 121 °C and 15 psi for 30 minutes. Radiation sterilized. DNase-, RNase-, Pyrogen-, and latex-free. Maximum RCF is 9000.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty