Would you like to log in again?
Micro Tube Rack, Polypropylene, 20-Place
Supplier: United Scientific Supplies
P20205
470339-526EA 81.3 USD InStock
470339-526
Micro Tube Rack, Polypropylene, 20-Place
Racks Tube Racks
Interlocking, autoclavable microcentrifuge tube rack.
  • Sturdy polypropylene racks hold 1.5 ml and 2.0 ml microcentrifuge tubes
  • Each rack features 20 places in a 10×2 array
  • Designed to lock side-by-side for larger experiments
  • Rack dimensions: 210L×70W×35H mm
  • Bottoms feature rubber supports for better stability
  • Autoclavable
Pack of 8 sturdy polypropylene racks, each able to hold up to 20 microcentrifuge tubes with 1.5 ml or 2.0 ml capacity. Racks are designed to lock side-by-side for larger experiments. Positions are numbered for easy identification of samples. Bottoms feature rubber supports for better stability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty