Would you like to log in again?
Clamp Holder, Right Angle
Supplier: United Scientific Supplies
RTCLMP1
470338-916EA 10.4 USD InStock
470338-916
Clamp Holder, Right Angle
Clamps General Purpose Clamps
Sturdy aluminum right angle clamp.
  • Sturdy aluminum design
  • Holds up to 21 mm rods
Cast aluminum clamp holder for use with rods up to 19 mm in diameter. For use when clamping at 90° is required. This clamp features a minimum to maximum grip range of 0 to 21 mm, and a v-groove bearing length of 55 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty