Would you like to log in again?
Equal Volume Density "Squares"
Supplier: United Scientific Supplies
SGVS04
470339-454EA 25.25 USD InStock
470339-454
Equal Volume Density "Squares"
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Square column metal set for density and specific gravity studies.
  • Set of 4 equal volume rectangular prisms
  • Includes: Aluminum, Copper, Steel and Brass
  • Dimensions: 1.26×1.26×5 cm (0.5×0.5×2")
  • Storage box and activity guide included
  • Ideal for lessons around density and specific gravity
Square specimen set is used for density and specific gravity studies. The square shape makes measurement and volume calculations easy for students. Set of four includes aluminum, copper, steel, and brass cylinders. Includes a storage box and activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty