Would you like to log in again?
Eco-Heat Hot Plate
Supplier: United Scientific Supplies
UNEHPLATE-1L
470339-542EA 181.55 USD InStock
470339-542
Eco-Heat Hot Plate
Heaters Hot Plates
An economical analog hotplate with stirrer that can heat up to 1000 ml of water.
  • Speed: 0 to 1600 rpm
  • Temperature range: 0 to 280 ℃
  • Maximum stirring capacity: 1 L
  • 12×12 cm aluminum top
This compact unit is ideal to warm small volumes of water to near boiling temperatures (~90 °C) in an open beaker. To boil water in an open beaker, we suggest selecting one of our advanced hot plate stirrers with higher surface temperatures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty