Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eco- Mini Centrifuge

Supplier: United Scientific Supplies
UNMCNTF-4
470338-936EA 202 USD InStock
470338-936
Eco- Mini Centrifuge
Centrifuges Microcentrifuges
A small centrifuge with lots of power.

  • MultiRotor: 12×0.2 ml/0.5 ml/1.5 ml/2 ml tubes
  • Strips-Rotor: 4×PCR strips (12×0.2 ml)
  • Maximum Speed: 4000 rpm
  • Maximum RCF: 1080×g

This bench-top microcentrifuge is ideal for quick spin-downs of microcentrifuge tubes. Includes a 12-position, 1.5/2 ml tube rotor as well as a 4-position, 12-strip PCR tube rotor that can accommodate 48 individual PCR tubes or 0.2 ml strip tubes.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty