Would you like to log in again?
Simple Resonance Tube
Supplier: United Scientific Supplies
UNRSTBKIT
470340-076EA 53.65 USD InStock
470340-076
Simple Resonance Tube
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Easy to use kit to investigate sound wavelength.
  • Designed for investigations of the relationship between wavelength, frequency, and speed of waves
  • Activity guide included
  • Enhance experiments with Smartphone apps
  • Correlates to NGSS Standards 4-PS4-1, MS-PS4-1, and HS-PS4-1
Explore the concept of acoustic resonance with this easy-to-use kit. Students will use a tuning fork or a smartphone app to generate sound through a metal tube of variable length, which is closed at one end and open at the other. They will investigate which lengths of tube amplify sound, drawing connections to the frequency and speed of the sound in air.
Contents: Telescoping metal tube of variable length (32 to 60 cm); Tape measure, 2 m; Tuning fork, 1024 Hz; Activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty