Would you like to log in again?
Basic Plant Anatomy Slide Set of 20
Supplier: United Scientific Supplies
UNSLPNT20
470339-264EA 47.85 USD InStock
470339-264
Basic Plant Anatomy Slide Set of 20
Slides Prepared Slides
Collection of popular plant samples.
  • Glass prepared microscope slides
  • Plastic storage case
  • Key chart is included on lid
Glass prepared microscope slides, supplied in a plastic case. A collection of slides specific to the study of plant anatomy. Set of 20.
Set includes: 1) Helianthus (sunflower), young, 2) Prunus (cherry), leaf, TS and old root, 3) Monocot, epidermis, E. for stomata, 4) Vicia (bean), young root, TS, 5) Vicia, root apex, LS, 6) Psamma (marram grass), leaf, TS, 7) Zea (maize), stem, TS and LS, 8) Lilium, anther, TS, 9) Cucurbita (marrow), stem, TS and LS, 10) Lilium, ovary, TS and LS, 11) Helianthus, stem, bundles, TS, 12) Lilium, ovary, various stages, TS, 13) Helianthus , old stem, TS and LS, 14) Capsella, mature cotyledon stage, LS, 15) Tilia (lime), young and old stem, TS, 16) Stigma, 17) Tilia, old stem, RLS, 18) Pollen grains, 19) Dicotyledon stem apex, LS, 20) Zea seed, LS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty