Would you like to log in again?
Water Sampler
Supplier: United Scientific Supplies
UNWTRSMPL01
470339-736EA 105.05 USD InStock
470339-736
Water Sampler
Samplers Water Samplers
Easy to use water sampler for depths up to 20 m.
  • Durable PVC construction
  • Activity guide included
  • Correlates to NGSS Standards MS-ESS3-3 and HS-ESS3-4
Capture water samples from specific depths with this clever tool. The unit is made from impact-resistant PVC, with soft rubber seals that hold up to 1.5 L of liquid. A unique design allows for hassle-free sample collection; when the desired depth is reached, students simply jerk the attached line to close the apparatus.
Includes a 20 m line on Styrofoam winding float. Unit is 30 cm long, 10 cm in diameter and weighs 2 pounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty