Would you like to log in again?
Unitary Wash Bottles with Colored Caps
Supplier: United Scientific Supplies
WBSET4
470339-772EA 22.65 USD InStock
470339-772
Unitary Wash Bottles with Colored Caps
Bottles Wash Bottles
Wash bottle with color coded caps.
  • Set of 4 wash bottles with 500 ml capacity and built-in delivery tube
  • Made of translucent, pliable low-density polyethylene (LDPE)
  • Dispense solutions easily with a light squeeze
  • Graduated on outer surface
Set of four wash bottles with 500 ml capacity and built-in delivery tube. Made of translucent, pliable low-density polyethylene (LDPE). Polypropylene caps included: one each of red, yellow, blue, and white.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty