Would you like to log in again?
Hooked Double-Ended Brass Weight Set of 9
Supplier: United Scientific Supplies
WHSET9
470339-964EA 140.5 USD InStock
470339-964
Hooked Double-Ended Brass Weight Set of 9
Weights Calibration Weights
Brass hooked weights with a storage block.
  • Can be used on balance pans, as well as hooked in tandem for lever arm studies
  • Sturdy storage block included
Set of nine weights consisting of one each 10, 50, 100, 500, 1000 g, and two each 20 g and 200 g weights. Weights feature flat countersunk bottoms, with a double-ended hooked rod through the middle. They can be used on balance pans, as well as hooked in tandem for lever arm studies. Weights are made of polished brass and come in a storage block.
Please note: Due to its size, the 10 g weight does not include a countersunk bottom.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty