Would you like to log in again?
Deluxe Slotted Weight Set of 12, with Rack
Supplier: United Scientific Supplies
WHST12
470340-250EA 129.5 USD InStock
470340-250
Deluxe Slotted Weight Set of 12, with Rack
Weights Calibration Weights
Slotted brass weights for use with hangers.
  • Set of 12 slotted brass weights
  • Durable rack also included
  • Also available as a set of 8 (WHST08) or 10 (WHST10)
Set of 12 high-quality slotted brass weights with durable rack.
Weights included: 1×500 g; 2×200 g; 1×100 g; 1×50 g; 2×20 g; 1×10 g; 1×5 g; 2×2 g; 1×1 g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty