Would you like to log in again?
Slotted Weight Set of 13, with Hanger and Case
Supplier: United Scientific Supplies
WHST13
470339-708EA 32.15 USD InStock
470339-708
Slotted Weight Set of 13, with Hanger and Case
Weights Calibration Weights
Slotten brass weights with an included hanger.
  • Set of 13 slotted weights made from brass
  • Includes hanger and case
Set of 13 slotted brass weights: 1×50 g, 9×20 g, 1×10 g, 2×5 g. Hanger and case also included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty