Would you like to log in again?
Scissors, Dissection, Sharp/Sharp, Straight, Mortech
Supplier: Mortech
AA101 AA002 AA102 AA003 AA103 AA001
76548-402EA 103.02 USD InStock
76548-402 76548-312 76548-406 76548-404 76548-308 76548-310
Scissors, Dissection, Sharp/Sharp, Straight, Mortech
Scissors
Sharp tipped scissors are designed for precise tissue cuts during dissection and surgical cases.
  • Stainless steel
  • Straight cutting edge
  • Sharp/sharp tips
Available in standard or premium grade.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty