Would you like to log in again?
Rib Shears, Councilman’s, Mortech
Supplier: Mortech
AF004
76549-176EA 320.58 USD InStock
76549-176
Rib Shears, Councilman’s, Mortech
Bone Cutters and Rongeurs
Councilman’s rib shears with box lock handle used for breaking or cutting bone. The long handles allow for maneuverability during rib cutting.
  • Standard grade stainless steel
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty