Would you like to log in again?
Bone Rongeurs, Cleveland, No. 4A, Mortech
Supplier: Mortech
AG034
76549-238EA 61.64 USD InStock
76549-238
Bone Rongeurs, Cleveland, No. 4A, Mortech
Bone Cutters and Rongeurs
Cleveland rongeurs are designed to cut, sculpt, and trim soft or hard bones. Satin surface to reduce glare.
  • German grade satin stainless steel
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty