Would you like to log in again?
Dissecting Board, Blue, Mortech
Supplier: Mortech
BC080
76549-610EA 152.03 USD InStock
76549-610
Dissecting Board, Blue, Mortech
Dissecting Boards
Manufactured from a high density polyethylene with a textured finish.
  • Manufactured from high density polyethylene with textured finish
  • Textured surface provides an excellent cutting surface
Bottom corners have a non-skid rubber foot attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty