Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Raised Body Boards, Mortech
Supplier: Mortech
BC081
76549-612EA 319.05 USD InStock
76549-612
Raised Body Boards, Mortech
Ergonomic Supports Back Supports
Body boards are used to raise the body above the work surface to allow for proper fluid drainage.
  • Machined from a single piece of high density polyethylene
  • Boards raise the body above the work surface allowing for added fluid drainage
  • Non-skid rubber feet are placed where contact is made with the work surface
  • Set of 3
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty