Would you like to log in again?
Mopec 1000 Autopsy Saw and Vacuum Set, Mortech
Supplier: Mortech
BD040+BD080
76549-616EA 7352.49 USD InStock
76549-616
Mopec 1000 Autopsy Saw and Vacuum Set, Mortech
Saws
This Mopec Autopsy Saw has a rugged construction and a machined aluminum housing that reduces heat and noise.
  • On/Off switch is designed and placed so that it will not break if dropped
Designed for removing the cranial cap, making linear cuts, or sectioning small bone specimens. The BD080 Bone Dust Vacuum Collector uses HEPA filtration to minimize exposure to particulate matter. The bone dust vacuum collector attaches directly to the autopsy saw for safety and efficiency.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty