Would you like to log in again?
Mopec 810 Saw and Vacuum Set, Mortech
Supplier: Mortech
BD810+BD080
76549-622EA 8956.39 USD InStock
76549-622
Mopec 810 Saw and Vacuum Set, Mortech
Saws
The Mopec 810 Autopsy Saw remains as the industry standard in anatomic and forensic pathology.
High speed oscillating action quickly cuts through bone with minimal damage to soft tissue. Used for removing the cranial cap, making linear cuts or sectioning small bone specimens.The BD080 Bone Dust Vacuum Collector uses HEPA filtration to minimize exposure to particulate matter. The bone dust vacuum collector attaches directly to the autopsy saw for safety and efficiency.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty