Would you like to log in again?
Splash Tubing, Mortech
Supplier: Mortech
BE017
76549-632EA 23.34 USD InStock
76549-632
Splash Tubing, Mortech
Tubing Transfer Tubing
Plastic Tubing that works with most morgue plumbing fixtures. Flexible for your needs.
  • Splash tubing for aspirator drain, embalming and autopsy tops
  • Sold in 3' sections
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty