Would you like to log in again?
ClearSteel, Stainless Steel Cleaner and Polish Spray, Mortech
Supplier: Mortech
ClearSteel
BE048
76549-636EA 68.01 USD InStock
76549-636
ClearSteel, Stainless Steel Cleaner and Polish Spray, Mortech
Cleaning Agents Detergents and Surface Cleansers
Powerful and non-corrosive cleaner for all stainless steel surfaces. The super concentrated biodegradable formula cleans and polishes in one step.
  • Easily remove fresh or dried blood and tissue
Ideal for autopsy tables, grossing stations and all stainless steel products including instruments, lighting, coolers, body trays and carriers. Use monthly to keep your lab clean and smelling fresh
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty