Would you like to log in again?
Disposable Poly Gown, BE300, Mortech
Supplier: Mortech
BE300
76549-686EA 3.49 USD InStock
76549-686
Disposable Poly Gown, BE300, Mortech
Disposable Garments
These level 1 disposable gowns are light and breathable.
  • One size fits most
  • Blue
  • 100% CPE
  • Over-the-head, open-back design with tie closure at waist
  • Long sleeves with thumb-hooks for comfort
  • Single-use, nonsterile
The over-the-head design makes them easy to put on and take off, while the long sleeves and thumb hooks ensure all-day comfort.
These gowns effectively protect against harmful germs, disease, and viruses. One-time use only. Dispose of immediately after use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty