Would you like to log in again?
Level IV Isolation Gown, Mortech
Supplier: Mortech
BE304 BE304XL
76549-688EA 11.44 USD InStock
76549-688 76549-690
Level IV Isolation Gown, Mortech
Disposable Garments
These level 4 isolation gowns offer the highest level of protection available and are used for high risk circumstances. The lightweight, breathable design allows for all-day comfort. They offer pathogen resistance, protection against non-airborne diseases, and are an effective barrier for fluids.
  • Level 4 Isolation
  • 75% SMS 25% PE
  • Blue
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty