Would you like to log in again?
RF Power Supplies for ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
8003-0560 8003-0560
AG8003-0560EA 1469 USD InStock
CAAG8003-0560 AG8003-0560
RF Power Supplies for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
RF load coils and power tubes for PerkinElmer NexION 300/350 and ELAN 9000/6x00/DRCs ICP-MS systems.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns, to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent radio frequency (RF) load coils and power tubes for PerkinElmer NexION 300/350 and ELAN 9000/6x00/DRCs and 5000 ICP-MS instruments.
Achieve guaranteed performance with supplies for PE from Agilent that are equivalent to the PerkinElmer ICP-MS versions so you can use supplies with complete confidence.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty