Would you like to log in again?
SPME Arrows, Agilent Technologies
Supplier: Agilent Technologies
5191-5861 5191-5867 5191-5860 5191-5859 5191-5868 5191-5858 5191-5866 5191-5868 5191-5869 5191-5858 5191-5860 5191-5864 5191-5862 5191-5869 5191-5867 5191-5865 5191-5859 5191-5866 5191-5861 5191-5865 5191-5862 5191-5864
AG5191-5868KT 3760 USD InStock
AG5191-5868 CAAG5191-5859 AG5191-5858 AG5191-5869 AG5191-5859 CAAG5191-5867 CAAG5191-5866 CAAG5191-5858 CAAG5191-5869 CAAG5191-5868 AG5191-5860 AG5191-5861 CAAG5191-5862 AG5191-5862 CAAG5191-5865 CAAG5191-5864 AG5191-5864 AG5191-5865 AG5191-5866 CAAG5191-5861 AG5191-5867 CAAG5191-5860
SPME Arrows, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
SPME Arrows are the next-generation improvement of solid phase microextraction fibers.
  • Available with a variety of polymer coatings
  • Up to 10 times more sensitivity than SPME fibers
SPME Arrows are the next-generation improvement of SPME fibers and can be used for headspace analysis or by immersion in a liquid to concentrate analyte on the Arrow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty