Would you like to log in again?
SPS 4 Update Kits for ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
G8410-60180 G8410-60180
AGG8410-60180KT 195 USD InStock
AGG8410-60180 CAAGG841060180
SPS 4 Update Kits for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A range of update kits for the SPS 4 extend sample handling and minimize contamination with ICP-MS.
  • Upgrade sample handling capacity, giving you flexibility to extend the capabilities of the SPS 4 autosampler used in your lab
  • A number of different autosamplers have been used with Agilent ICP-MS instruments, depending on the specific model
  • The SPS 4 is used with current instruments
  • The autosampler is designed to automate the presentation of liquid samples, drawn from tubes held in racks loaded on the rack location mat, to the instrument for analysis
  • Each autosampler can accommodate a variety of fixtures, including standards and sample racks, probes and other accessories
Agilent offers a range of update kits and optional accessories to upgrade sample handling capacity with the Agilent SPS 4 when used with Agilent ICP-MS instruments.
These kits include high volume and pumped rinse kits, dual rinse stations and autosampler covers with extraction designed to minimize risk of environmental contamination of trace samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty