Would you like to log in again?
Bond Elut PSA, Agilent Technologies
Supplier: Agilent Technologies
12256140 12256140 12256055 12256055
AG12256140PK 216 USD InStock
CAAG12256055 CAAG12256140 AG12256140 AG12256055
Bond Elut PSA, Agilent Technologies
Chromatography Columns
Bond Elut PSA is an alkylated amine with two different amino functionalities.
  • Alternative choice to Bond Elut NH2 for polar compounds
  • Higher ionic capacity than NH2
Bond Elut PSA is an alkylated amine sorbent that contains two different amino functionalities. This gives a slightly higher pKa and ionic capacity compared to Bond Elut NH2. PSA has a significantly higher carbon load than most amino functional sorbents and is a good choice for polar compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty