Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium bromide for IR spectrophotometry, Fluka™

Supplier: Honeywell Research Chemicals

34919-6X100G
BJ34919-100GCS 2325.46 USD InStock
BJ34919-100G
Potassium bromide for IR spectrophotometry, Fluka™
Potassium bromide


Formula: KBr
MW: 119 g/mol
Boiling Pt: 1.383 °C (1.013 hPa)
Melting Pt: 734 °C
Density: 2.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Order Now
Specification Test Results
Suitability for IR spectroscopy complying
Vapor Pressure <0.01 mmHg ( 20 °C)
Vapor Pressure 1 mmHg ( 795 °C)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty