Would you like to log in again?
Functional Eye Model
Supplier: ARCO
4072800
470346-236EA 51.45 USD InStock
470346-236
Functional Eye Model
Models Human Biology Models
Examine how the eye works.
  • Adjustable lens curvature
  • Viewing object
  • Projection panel
A unique apparatus allows you to recreate the conditions of visual deficiencies in the human eye. The clear silicone elastic lens chamber of the model is filled with water. When water is added or subtracted with the included syringe, the curvature of the lens is changed, which alters the focus of images projected on the retina. The shape of the eyeball is adjustable as well. Once the deficiencies have been observed, the included corrective lenses can be added to show how vision defects can be modified. The five-piece model is mounted on a base with the projection object on a separate stand. The set comes with two lenses, lens holder, projection object, model, and instruction manual with numbered Key Card.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty