Would you like to log in again?
TraxIt® Single Use Thermometers
Supplier: MEDICAL INDICATORS,INC
1313-20
470346-244BX 76.29 USD InStock
470346-244
TraxIt® Single Use Thermometers
Thermometers Digital Thermometers
The ideal thermometer for pediatric care.
  • Single patient use
  • 48 hour continuous read
TraxIt® thermometers are meant to simplify the process of taking a child’s temperature. Instead of having to repeatedly take a child’s temperature, the TraxIt® adheres to the child’s underarm and continuously reads the child’s temperature for up to 48 hours. TraxIt®’s adhesive is a gentle, latex-free, waterproof, hypoallergenic medical tape that will not irritate a child’s healthy skin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty