Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride dihydrate 99-103%, puriss., meets analytical specification of FCC, Ph. Eur., USP, E509 (max. 0.0001% Al), Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Calcium Chloride Dihydrate

12022-6X2.5KG
BJ12022-2.5KGCS 2063.85 USD InStock
BJ12022-2.5KG
Calcium chloride dihydrate 99-103%, puriss., meets analytical specification of FCC, Ph. Eur., USP, E509 (max. 0.0001% Al), Fluka™
Calcium chloride dihydrate


Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Order Now
Specification Test Results
Assay 99.0 - 103.0 %
pH (5 %, 25 °C) 4.5 - 8.5
Free acid (as HCl) Max. 0.005 %
Acidity or alkalinity complying
Free alkali (as CaO) Max. 0.005 %
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 2 ppm
Barium (Ba) complying
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.05 %
Sodium (Na) Max. 0.02 %
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 10 ppm
Fluoride (F) Max. 40 ppm
Sulfate (SO4) Max. 200 ppm
Aluminium, iron and phosphates complying
Mg and alkali salts (SO4) Max. 0.5 %
Residual solvents complying
Appearance of the solution complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty