Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accu-CT Split-Core Current Transformer 333mV Sensor, Onset Computer Corporation

Supplier: Onset
T-ACT-0750-020 T-ACT-0750-250 T-ACT-0750-050 T-ACT-0750-100
470346-432EA 124.89 USD InStock
470346-432 470346-450 470346-428 470346-438
Accu-CT Split-Core Current Transformer 333mV Sensor, Onset Computer Corporation
Data Loggers
The Accu-CT split-core current transformer offers outstanding accuracy and one-handed operation.

  • Unprecedented Linearity
  • Window Opening 0.75 Inch
  • The Accu-CT revenue grade, split-core current transformer offers outstanding linearity and phase angle accuracy with a unique one-handed opening and closing mechanism
  • Safe: burden resistor built-in, 333 mVac voltage output at rated full scale current, no shorting blocks needed
  • IEEE C57.13 and IEC 60044-1 accuracy: over full temperature range and down to 1% of rated current
  • Exceptionally low phase angle error: essential for accurate power and energy measurements
  • Glove-friendly: easy to open and install with one hand while wearing safety gloves

Certifications: UL recognized, CE mark, RoHS

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty