Would you like to log in again?
Innovating Science® Replacement Urine for IS3008
Supplier: ALDON CORP SE
IS3008-REF
470343-480EA 46.1 USD InStock
470343-480
Innovating Science® Replacement Urine for IS3008
Educational Classroom Kits and Activities
Learn about urinalysis, one of the oldest medical diagnostic tests performed.
  • Students will use simulated urine to test factors like pH, color, clarity, and test for the presence of factors such as proteins, glucose, and calcium
  • Aligned to NGSS Practices
Urinalysis, one of the oldest medical diagnostic tests performed, is to this day still one of the most common. In this activity, students will use simulated urine to avoid the unpleasantness of using the real thing while still performing actual tests used on real urine samples. Students will examine the simulated urine for factors such as pH, color, clarity, as well as test for the presence or absence of proteins, glucose, and calcium.
The students will then examine the samples microscopically to determine if crystals may be present in any of the samples. Kit contains enough materials for 15 groups of students. Teacher's Manual and Student Study Guide copymasters are included. Needed but not supplied are a hot water bath, glass test tubes, and compound microscopes (4X/10X/40X).
Delivery information: Kit includes instructional manual and SDS booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty