Would you like to log in again?
Innovating Science® Cellular Respiration - What Sugar does Yeast Like Best?
Supplier: ALDON CORP SE
Innovating Science®
IS1510 IS3012
470343-488EA 44.9 USD InStock
470343-488 470330-514
Innovating Science® Cellular Respiration - What Sugar does Yeast Like Best?
Educational Classroom Kits and Activities
Learn about how yeast may use different options for energy production.
  • Students will expose living yeast cells to three potential food sources and determine the effectiveness of the food sources
  • Aligned to NGSS Practices
  • Performance expectations: HS-LS1-7, HS-LS2-3
  • Cross cutting concepts include energy and matter
  • Engineering practices include developing and using models, planning and carrying out investigations
In this experiment you will expose living yeast cells to three different sugars. The sugars used will be glucose, sucrose, and lactose. When living yeast cells are exposed to these sugars, the cells should begin to utilize the sugars as a food source if they are capable of metabolizing them. Upon using the sugars, the yeast cells will begin to engage in aerobic respiration and/or fermentation.
This lab allows students to understand that yeast may use different options for energy production. Students will expose living yeast cells to three different potential food sources and use a pH indicator to indirectly determine the effectiveness of three different sugars as a food source for yeast. Kit contains enough materials for 15 groups.
Delivery information: Instructor's Manual and SDS included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty