Would you like to log in again?
Innovating Science® Testing Food for Nutrients
Supplier: ALDON CORP SE
IS3013
470343-490EA 73.65 USD InStock
470343-490
Innovating Science® Testing Food for Nutrients
Educational Classroom Kits and Activities
Learn about the nutrients in food including proteins, carbohydrates, and lipids.
  • Students will test for proteins, sugar, starch, and lipids to identify food nutrients
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
This experiment will help students understand the importance of proteins, carbohydrates, and lipids in living organisms. They will learn to identify a positive test result for proteins using biuret reagent and examine the reaction between Benedict's reagent and a simple sugar. Using iodine/potassium iodide they will test for the presence of starch and test for the presence of lipids using a fat-soluble dye.
Delivery information: Kit includes instructional manual and SDS booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty