Would you like to log in again?
Innovating Science® Chemiluminescence in Blood Stain Detection
Supplier: ALDON CORP SE
IS9009
470343-406EA 38.4 USD InStock
470343-406
Innovating Science® Chemiluminescence in Blood Stain Detection
Educational Classroom Kits and Activities
Learn about how crime scene investigators use chemiluminescence to test for blood stains.
  • Students will observe how using a special luminol formulation detect blood stains that are naked to the human eye.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data
Crime scene investigators are called to the scene of a possible violent crime. They examine the scene for evidence, such as fingerprints, hair, fibers, etc. After collecting the evidence, they notice there is no visible blood. Someone takes out a spray bottle and begins to spray the area with a liquid. After the area is covered with spray, they turn out the lights. A strange, faint glow appears in certain areas of the scene.
Learn how luminol is used in scenes like this everyday. The special luminol formulation does not require a separate hydrogen peroxide catalyst. Simply re-hydrate and use with the simulated blood hemoglobin to show your class. This activity includes an Instruction Manual with suggested activities. Kit contains enough material for several demonstrations.
Delivery information: Kit includes instructional manual and SDS booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty