Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

HYDRANAL™
37863-6X1L
BJ37863-1LCS 464.15 USD InStock
BJ37863-1L
Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 60-62 °C (1.013 hPa)
Melting Pt: -63 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 3,III
REACH: 05-2114614906-45
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Suitability (for KF-titration) test passed
Assay of water Max. 0.01 %
Number Of Samples Min. 2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty