Would you like to log in again?
Innovating Science® Separation of a Mixture of Solids
Supplier: ALDON CORP SE
Innovating Science®
IS2523-SGL IS2523
470231-550KT 38.55 USD InStock
470231-550 470343-452
Innovating Science® Separation of a Mixture of Solids
Educational Classroom Kits and Activities
Learn about how the properties of solubility can be utilized to separate 2 solids in a mixure.
  • Students will perform a qualitative test that will show how 2 different solids were present in one mixture, and how to separate them.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-1; MS-PS1-2; MS-PS1-5; HS-PS1-2; HS-PS1-3; HS-PS1-7
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Scale proportion and quantity, patterns, energy and matter, scientific knowledge assumes an order and consistency in natural systems
  • Engineering practices include eveloping and Using Models; Analyzing and Interpreting Data; Planning and Carrying Out Investigations; Scientific Knowledge is Based on Empirical Evidence; Science Models, Laws, Mechanisms, and theories Explain Natural Phenomena; Constructing Explanations and Designing Solutions; Using Mathematics and Computational Thinking
Performing liquid-liquid extractions allows students to apply their knowledge of the properties of molecules. Students will learn how the properties of solubility can be utilized to separate two solids in a mixture. This kit provides for a qualitative test that will help students visualize that two different solids were collected, despite their similar appearance. Students will also explore the concept of the law of conservation of mass as well as how acid-base reactions can be utilized to manipulate molecules. This kit contains enough material for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Instructor's Manual and SDS included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty