Would you like to log in again?
United Scientific™ Demonstration Lens Set
Supplier: United Scientific Supplies
DEM038 DEM075 DEM050
470339-424EA 13.5 USD InStock
470339-424 470340-118 470339-956
United Scientific™ Demonstration Lens Set
Educational Materials Physics Educational Materials Optics Learning Activities
Introductory lens set for optics experiments.
  • Concave and convex selection
  • All focal lengths are 150 mm
The set contains six 38 mm lenses, one each of double convex, plano-convex,concavo-convex, double concave, plano-concave, and convexo-concave lens. Comes in a reusable cardboard storage box. Lenses in all sets have focal lengths on 150 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty