Would you like to log in again?
Cole-Parmer® Essentials Critical Task Wipes, Antylia Scientific
Supplier: Antylia Scientific
78902-93 78902-95
76551-210BX 5.6 USD InStock
76551-210 76551-212
Cole-Parmer® Essentials Critical Task Wipes, Antylia Scientific
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
Made from 100% recycled cellulose (50% post consumer), these environmentally friendly lab wipes are excellent for light-duty applications, will not scratch, and can be used in a variety of industries.
  • Ideal for optics and other sensitive surfaces
  • Perfect wipe for light-duty tasks and delicate surfaces
The wipes are made of single-ply, absorbent, and soft material making them ideal for cleaning small parts, lenses, and other equipment in lab facilities. Since they are low linting and low in chemical extractables, they will leave surfaces extremely clean. The wipes are a complete ecologically friendly wipe option.
Packaging: Packaged in recycled and biodegradable boxes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty