Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: m-Bromobenzonitrile

10670-1000 10670-0100 10670-0025
TS10670-1000EA 398 USD InStock
TS10670-1000 TS10670-0025 TS10670-0100
3-Bromobenzonitrile 99%
3-Bromobenzonitrile


Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 225 °C (760 mmHg)
Melting Pt: 37…41 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001796
CAS Number: 6952-59-6
EINECS: 230-127-9
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to light yellow to beige Crystalline powder or crystals or crystalline mass
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty