Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-chlorobenzene 98.5%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromochlorobenzene

10682-1000 10682-0250
TS10682-1000EA 108 USD InStock
TS10682-1000 TS10682-0250
1-Bromo-2-chlorobenzene 98.5%
1-Bromo-2-chlorobenzene


Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 204 °C (760 mmHg)
Density: 1.649 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00000532
CAS Number: 694-80-4
EINECS: 211-775-1
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light orange Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5805 to 1.5825 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty