Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorobenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (4-Chlorophenyl)methanamine

10879-0025 10879-0500 10879-1000 10879-0250
TS10879-1000EA 226 USD InStock
TS10879-1000 TS10879-0025 TS10879-0500 TS10879-0250
4-Chlorobenzylamine 98%
4-Chlorobenzylamine


Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 215 °C (760 mmHg)
Density: 1.16 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008121
CAS Number: 104-86-9
EINECS: 203-245-3
UN: 2922
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5576 to 1.5610 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤50 APHA

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty