Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dibromo-2-nitrobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,5-Dibromonitrobenzene

11286-1000 11286-0250
TS11286-1000EA 151 USD InStock
TS11286-1000 TS11286-0250
1,4-Dibromo-2-nitrobenzene 97%
1,4-Dibromo-2-nitrobenzene


Formula: C₆H₃Br₂NO₂
MW: 280.9 g/mol
Melting Pt: 81…85 °C
Density: 2.374 g/cm³
MDL Number: MFCD00007046
CAS Number: 3460-18-2
EINECS: 222-404-8
UN: 2811
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 85 °C
GC ≥96.0 %
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty