Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichlorobenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (2,4-Dichlorophenyl)methanamine

11323-0100 11323-0500
TS11323-0100EA 68 USD InStock
TS11323-0100 TS11323-0500
2,4-Dichlorobenzylamine 98%
2,4-Dichlorobenzylamine


Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 83 °C (1013 hPa)
Density: 1.32 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00008109
CAS Number: 95-00-1
EINECS: 202-382-6
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5760 to 1.5800 (20°C, 589 nm)
GC ≥97.5 %
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥77000 l/(mol.cm) (at Lambda max)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty