Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibenzosuberone 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Dibenzosuberone1-Dibenzo[b,f]cycloheptanoneDibenzo[b,f]cycloheptan-1-one10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one

11474-1000
TS11474-1000EA 85 USD InStock
TS11474-1000
Dibenzosuberone 97%
Dibenzosuberone


Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 148 °C (.3 mmHg)
Melting Pt: 32…34 °C
Density: 1.15 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003587
CAS Number: 1210-35-1
EINECS: 214-912-3
Order Now
Specification Test Results
Appearance Clear yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
Titration with NaOH ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty