Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dimethoxybenzyl alcohol 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3,4-Dimethoxyphenyl)methanolVeratryl alcohol

11549-0250 11549-5000 11549-1000
TS11549-0250EA 47 USD InStock
TS11549-0250 TS11549-5000 TS11549-1000
3,4-Dimethoxybenzyl alcohol 96%
3,4-Dimethoxybenzyl alcohol


Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 296…297 °C (732 mmHg)
Melting Pt: 22 °C
Density: 1.15 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004638
CAS Number: 93-03-8
EINECS: 202-212-0
Order Now
Specification Test Results
Appearance Clear colorless to yellow to golden Viscous liquid
Infrared spectrum Conforms
GC ≥95 %
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty