Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dinitroaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

11693-0050 11693-5000 11693-1000
TS11693-5000EA 298 USD InStock
TS11693-5000 TS11693-0050 TS11693-1000
2,4-Dinitroaniline 99%
2,4-Dinitroaniline


Formula: C₆H₅N₃O₄
MW: 183.12 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 177…180 °C
Flash Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00007151
CAS Number: 97-02-9
EINECS: 202-553-5
UN: 1596
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Yellow to yellow-green to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 176 to 180 °C
Water 8 to 15 % (K.F.)
HPLC ≥98.5 %
Copper (Cu) ≤20 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤50 ppm
Nickel (Ni) ≤1000 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤300 ppm
Insoluble matter ≤0.01 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty