Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Fluorobenzenethiol4-Fluorobenzene-1-thiol1-Fluoro-4-mercaptobenzenep-Fluorothiophenol

11965-0100 11965-0500
TS11965-0500EA 251 USD InStock
TS11965-0500 TS11965-0100
4-Fluorothiophenol 97%
4-Fluorothiophenol


Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 164 °C (1013 hPa)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00004846
CAS Number: 371-42-6
EINECS: 206-737-6
UN: 1228
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5490 to 1.5520 (20°C, 589 nm)
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty